missing translation for 'onlineSavingsMsg'
Learn More

(1-Benzyl-4-piperidyl)methanol, Thermo Scientific™, Thermo Scientific™

Product Code. 10061194
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10061194 5 g 5g
10561132 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061194 Supplier Thermo Scientific Maybridge Supplier No. KM09623DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67686-01-5
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD00831014
InChI Key FLQPYEOKVZYXRL-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidyl methanol, 1-benzylpiperidin-4-yl methanol, 1-benzyl-4-piperidinemethanol, 1-benzyl-4-piperidinyl methanol, 1-benzyl-4-hydroxymethylpiperidine, 4-piperidinemethanol, 1-phenylmethyl, 1-benzy-4-hydroxymethylpiperidine, 1-benzy-4-hydroxymethyl piperidine, pubchem8894, 1-benzyl-4-piperdinemethanol
PubChem CID 736802
IUPAC Name (1-benzylpiperidin-4-yl)methanol
SMILES C1CN(CCC1CO)CC2=CC=CC=C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 205.3
Percent Purity 97%
Chemical Name or Material (1-Benzyl-4-piperidyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.