missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-piperidone, 98+%

Product Code. 11439553
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439553 100 g 100g
11429553 25 g 25g
11449553 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439553 Supplier Thermo Scientific Alfa Aesar Supplier No. A12390.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3612-20-2 | C12H15NO | 189.258 g/mol

1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

Solubility
Soluble in water (12 g/100ml at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

Density 1.059
Boiling Point 248°C
Flash Point >110°C (230°F)
Refractive Index 1.5399
Quantity 100 g
Beilstein 128556
Solubility Information Soluble in water (12g/100ml at 20°C).
Formula Weight 189.26
Percent Purity ≥98%
Chemical Name or Material 1-Benzyl-4-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.