missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-cyano-4-hydroxypiperidine, 98%, Thermo Scientific™

Product Code. 10098551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10098551 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10098551 Supplier Thermo Scientific Acros Supplier No. 136860100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6094-60-6
Molecular Formula C13H16N2O
Molecular Weight (g/mol) 216.28
MDL Number MFCD00023752
InChI Key XQRLXUYZKZXSBN-UHFFFAOYSA-N
Synonym 1-benzyl-4-cyano-4-hydroxypiperidine, maybridge1_006722, acmc-20aoxx, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
PubChem CID 80189
IUPAC Name 1-benzyl-4-hydroxypiperidine-4-carbonitrile
SMILES OC1(CCN(CC2=CC=CC=C2)CC1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95°C to 102°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 216.28
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Benzyl-4-cyano-4-hydroxypiperidine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.