missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrroline, 98%

Product Code. 11318856
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318856 5 g 5g
11308856 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318856 Supplier Thermo Scientific Alfa Aesar Supplier No. L19496.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6913-92-4 | C11H13N | 159.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6913-92-4
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00012324
InChI Key LRFHKHHUKGZIGE-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
PubChem CID 561506
IUPAC Name 1-benzyl-2,5-dihydropyrrole
SMILES C1C=CCN1CC2=CC=CC=C2

Specifications

Density 0.837
Boiling Point 60°C (1 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.542
Quantity 5 g
UN Number UN2735
Beilstein 111101
Sensitivity Air sensitive
Formula Weight 159.23
Percent Purity ≥98%
Chemical Name or Material 1-Benzyl-3-pyrroline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.