missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrroline, 97%, Thermo Scientific Chemicals

Product Code. 11926651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11926651 1 g 1g
11936651 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926651 Supplier Thermo Scientific Acros Supplier No. 436770010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6913-92-4
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
InChI Key LRFHKHHUKGZIGE-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
PubChem CID 561506
IUPAC Name 1-benzyl-2,5-dihydropyrrole
SMILES C1C=CCN1CC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8370g/mL
Boiling Point 92°C (1.0 hPa)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 0.837
Formula Weight 159.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-pyrroline
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzyl-3-pyrroline
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.