missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole 97+%, Thermo Scientific™

Product Code. p-4510314
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182834 250 mg Amber Glass Bottle 250mg
10684195 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182834 Supplier Thermo Scientific Maybridge Supplier No. MO07869CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 214540-43-9
Molecular Formula C9H7BrFN3
Molecular Weight (g/mol) 256.078
InChI Key MUYAKFNLCJMPDH-UHFFFAOYSA-N
Synonym 1-benzyl-3-bromo-5-fluoro-1h-1,2,4-triazole, 1h-1,2,4-triazole,3-bromo-5-fluoro-1-phenylmethyl, 3-bromo-5-fluoro-1-benzyl-1,2,4-triazole, 1-benzyl-3-bromo-5-fluoro-1h-1,2,4 triazole, 3-bromo-5-fluoro-1-phenylmethyl-1,2,4-triazole, 3-bromanyl-5-fluoranyl-1-phenylmethyl-1,2,4-triazole
PubChem CID 10901231
IUPAC Name 1-benzyl-3-bromo-5-fluoro-1,2,4-triazole
SMILES C1=CC=C(C=C1)CN2C(=NC(=N2)Br)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 256.08
Percent Purity 97%
Chemical Name or Material 1-Benzyl-3-bromo-5-fluoro-1H-1, 2, 4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.