missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 256.08 |
Percent Purity | 97% |
Chemical Name or Material | 1-Benzyl-3-bromo-5-fluoro-1H-1, 2, 4-triazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'