missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-benzyl-3,5-dibromo-1H-1,2,4-triazole, Thermo Scientific™
Click to view available options
1g
Specifications
CAS | 106724-85-0 |
Assay Percent Range | 95% |
Molecular Formula | C9H7Br2N3 |
Quantity | 1 g |
Synonym | 1-benzyl-3,5-dibromo-1h-1,2,4-triazole, benzyldibromotriazole, acmc-1buio, 3,5-dibromo-1-benzyl-1,2,4-triazole, 3,5-dibromo-1-phenylmethyl-1,2,4-triazole, 1-benzyl-3,5-dibromo-1h-1,2,4 triazole, 1h-1,2,4-triazole, 3,5-dibromo-1-phenylmethyl, 3,5-bis bromanyl-1-phenylmethyl-1,2,4-triazole |
InChI Key | YXSNCHAOAYRKCF-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)CN2C(=NC(=N2)Br)Br |
IUPAC Name | 1-benzyl-3,5-dibromo-1,2,4-triazole |
Molecular Weight (g/mol) | 316.984 |
PubChem CID | 1475046 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'