missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-pyrrolidinone, 97%

Product Code. 10269593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269593 500 g Glass bottle 500g
10091770 25 g Glass bottle 25g
10591591 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269593 Supplier Thermo Scientific Acros Supplier No. 154125000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5291-77-0 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5291-77-0
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00003195
InChI Key LVUQCTGSDJLWCE-UHFFFAOYSA-N
Synonym 1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
PubChem CID 79176
IUPAC Name 1-benzylpyrrolidin-2-one
SMILES C1CC(=O)N(C1)CC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5500 to 1.553
Quantity 500 g
Specific Gravity 1.09
Formula Weight 175.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-2-pyrrolidinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.