missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-1H-indol-5-ylamine, 97%, Thermo Scientific™

Catalog Number 10416862
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
26807-73-8
C15H14N2
222.291
MFCD03070173
UYDNPZLYDODKKA-UHFFFAOYSA-N
1-benzyl-1h-indol-5-ylamine, 1-benzyl-1h-indol-5-amine, 5-amino-1-benzylindole, n-benzyl-5-amino indole, 1-benzylindole-5-ylamine, 5-amino-1benzyl-1h-indole, 5-amino-1-benzyl-1h-indole, 1h-indol-5-amine,1-phenylmethyl
2794624
1-benzylindol-5-amine
C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)N
This item is not returnable. View return policy
26807-73-8
C15H14N2
222.291
MFCD03070173
UYDNPZLYDODKKA-UHFFFAOYSA-N
1-benzyl-1h-indol-5-ylamine, 1-benzyl-1h-indol-5-amine, 5-amino-1-benzylindole, n-benzyl-5-amino indole, 1-benzylindole-5-ylamine, 5-amino-1benzyl-1h-indole, 5-amino-1-benzyl-1h-indole, 1h-indol-5-amine,1-phenylmethyl
2794624
1-benzylindol-5-amine
C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'