missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-1,4,7,10-tetraazacyclododecane

Product Code. 15453487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453487 250 mg 250mg
11465930 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453487 Supplier Thermo Scientific Alfa Aesar Supplier No. H26525.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112193-83-6 | C15H26N4 | 262.401 g/mol

Used as ligands for catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligands for catalysts.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112193-83-6
Molecular Formula C15H26N4
Molecular Weight (g/mol) 262.401
MDL Number MFCD09263310
InChI Key FURLCQRFFWBENR-UHFFFAOYSA-N
Synonym 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min
PubChem CID 10084082
IUPAC Name 1-benzyl-1,4,7,10-tetrazacyclododecane
SMILES C1CNCCN(CCNCCN1)CC2=CC=CC=C2

Spécification

Melting Point 84°C to 86°C
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 262.4
Chemical Name or Material 1-Benzyl-1,4,7,10-tetraazacyclododecane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.