missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylpyrene, 98%, Thermo Scientific™

Product Code. 15187432 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15187432

Brand: Thermo Scientific Acros 449310010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 127.0°C to 129.0°C
CAS 7376-03-6
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C10H11N2O3
Packaging Glass Bottle
Synonym n-4-aminobenzoyl-beta-alanine, 3-4-amino-benzoylamino-propionic acid, 3-4-aminobenzamido propanoic acid, 4-aminobenzoyl-beta-alanine, unii-ab85ljq945, 3-4-aminobenzoyl amino propanoic acid, n-4-aminophenyl carbonyl-beta-alanine, 3-4-aminophenyl formamido propanoic acid, 3-4-aminophenyl carbonylamino propanoic acid
Solubility Information Solubility in water: insoluble
InChI Key VHAXWROFYVPXMZ-UHFFFAOYSA-M
SMILES NC1=CC=C(C=C1)C(=O)NCCC([O-])=O
IUPAC Name 3-[(4-aminobenzoyl)amino]propanoic acid
Molecular Weight (g/mol) 207.21
Quantity 1 g
PubChem CID 719629
Formula Weight 306.36
Percent Purity 98%
Chemical Name or Material 1-Benzoylpyrene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.