missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylacetone, 98%

Product Code. 10569620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10569620 250 g Glass bottle 250g
10427670 50 g Glass bottle 50g
11984841 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10569620 Supplier Thermo Scientific Acros Supplier No. 105692500
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-91-4 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Yellow
Density 1.0900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5COCH2COCH3
Quantity 250 g
Beilstein 07,68
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Flakes or Crystalline Powder
Chemical Name or Material 1-Benzoylacetone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.