missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylacetone, 98%, Thermo Scientific Chemicals

Catalog Number 10427670
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
93-91-4
C10H10O2
162.19
MFCD00008786
CVBUKMMMRLOKQR-UHFFFAOYSA-N
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
7166
1-phenylbutane-1,3-dione
CC(=O)CC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
93-91-4
C10H10O2
162.19
MFCD00008786
CVBUKMMMRLOKQR-UHFFFAOYSA-N
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
7166
1-phenylbutane-1,3-dione
CC(=O)CC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Yellow
Density 1.0900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5COCH2COCH3
Quantity 50 g
Beilstein 07,68
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Flakes or Crystalline Powder
Chemical Name or Material 1-Benzoylacetone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'