missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10132984
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132984 5 g Amber Glass Bottle 5g
10065972 250 mg Amber Glass Bottle 250mg
10542423 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10132984 Supplier Thermo Scientific Maybridge Supplier No. CC24804DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10133-30-9
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD05663673
InChI Key QHHRWAPVYHRAJA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde
PubChem CID 139097
IUPAC Name 1-benzothiophene-5-carbaldehyde
SMILES O=CC1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 162.21
Percent Purity 95%
Chemical Name or Material 1-Benzothiophene-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.