missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-3-ylmethanol, 97%, Thermo Scientific™

Product Code. 10215224
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10215224 10 g 10g
10581682 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215224 Supplier Thermo Scientific Maybridge Supplier No. CC12309EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5381-24-8
Molecular Formula C9H8OS
Molecular Weight (g/mol) 164.22
MDL Number MFCD02682007
InChI Key UYGMKSKKGSUAHB-UHFFFAOYSA-N
Synonym benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene
PubChem CID 2776342
SMILES OCC1=CSC2=CC=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name (1-benzothiophen-3-yl)methanol
Formula Weight 164.23
Percent Purity 97%
Chemical Name or Material 1-Benzothiophen-3-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.