missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylazetidine-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 15491929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491929 5 g 5g
15401939 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491929 Supplier Thermo Scientific Alfa Aesar Supplier No. H64647.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36476-86-5
Molecular Formula C17H16N2
Molecular Weight (g/mol) 248.33
MDL Number MFCD00158910
InChI Key IXMOEAHDRKNAAG-UHFFFAOYSA-N
Synonym 1-benzhydrylazetane-3-carbonitrile, 1-benzhydryl-3-cyanoazetidine, 1-benzhydryl-azetidine-3-carbonitrile, 1-diphenylmethyl azetidine-3-carbonitrile, 3-cyano-1-diphenylmethyl azetidine, 3-azetidinecarbonitrile, 1-diphenylmethyl, 1-1,1-diphenylmethyl azetidine-3-carbonitrile, 1-diphenylmethyl-3-cyanoazetidine, 1-diphenylmethyl-3-azetidinecarbonitrile, 1-benzhydryl-3-azetanecarbonitrile
PubChem CID 2779290
SMILES N#CC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 150°C to 153°C
Quantity 5 g
UN Number UN3439
Beilstein 482511
IUPAC Name 1-(diphenylmethyl)azetidine-3-carbonitrile
Formula Weight 248.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzhydrylazetidine-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.