missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylazetane-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 10222842
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10222842 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10222842 Supplier Thermo Scientific Maybridge Supplier No. KM08760DA
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 36476-86-5
Assay Percent Range 97%
Molecular Formula C17H16N2
MDL Number MFCD00158910
Quantity 1 g
Synonym 1-benzhydrylazetane-3-carbonitrile, 1-benzhydryl-3-cyanoazetidine, 1-benzhydryl-azetidine-3-carbonitrile, 1-diphenylmethyl azetidine-3-carbonitrile, 3-cyano-1-diphenylmethyl azetidine, 3-azetidinecarbonitrile, 1-diphenylmethyl, 1-1,1-diphenylmethyl azetidine-3-carbonitrile, 1-diphenylmethyl-3-cyanoazetidine, 1-diphenylmethyl-3-azetidinecarbonitrile, 1-benzhydryl-3-azetanecarbonitrile
InChI Key IXMOEAHDRKNAAG-UHFFFAOYSA-N
SMILES N#CC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 1-benzhydrylazetidine-3-carbonitrile
Molecular Weight (g/mol) 248.33
PubChem CID 2779290
Formula Weight 248.33
Chemical Name or Material 1-Benzhydrylazetane-3-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.