missing translation for 'onlineSavingsMsg'
Learn More

1-Azepanyl(4-piperidinyl)methanone, ≥97%, Thermo Scientific™

Product Code. 10377294
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10377294 10 g 10g
10300853 250 mg 250mg
10463583 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377294 Supplier Thermo Scientific Maybridge Supplier No. MO01173EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 86542-89-4
Molecular Formula C12H22N2O
Molecular Weight (g/mol) 210.321
MDL Number MFCD03372514
InChI Key BWKOHTHENZSVRL-UHFFFAOYSA-N
Synonym 1-piperidin-4-ylcarbonyl azepane, 1-azepanyl 4-piperidinyl methanone, azepan-1-yl-piperidin-4-yl-methanone, azepan-1-yl piperidin-4-yl methanone, 1-piperidine-4-carbonyl azepane, methanone, hexahydro-1h-azepin-1-yl-4-piperidinyl, azaperhydroepinyl 4-piperidyl ketone, 1-4-piperidylcarbonyl azepane, 1-piperid-4-ylcarbonyl azepane
PubChem CID 2794691
IUPAC Name azepan-1-yl(piperidin-4-yl)methanone
SMILES C1CCCN(CC1)C(=O)C2CCNCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 210.32
Percent Purity 97%
Chemical Name or Material 1-Azepanyl(4-piperidinyl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.