missing translation for 'onlineSavingsMsg'
Learn More

1-Arachidoyl-sn-glycero-3-phosphocholine, MedChemExpress

Product Code. 16404748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16404748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16404748 Supplier Medchem Express Supplier No. HY113010
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 1-Arachidoyl-sn-glycero-3-phosphocholine is a lysophospholipid (LyP).

1-Arachidoyl-sn-glycero-3-phosphocholine is a lysophospholipid (LyP).

Specifications

Chemical Name or Material 1-Arachidoyl-sn-glycero-3-phosphocholine
Physical Form Solid
CAS 108341-80-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H58NO7P
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES CCCCCCCCCCCCCCCCCCCC(OC[C@@H](O)COP(OCC[N+](C)(C)C)([O-])=O)=O
Molecular Weight (g/mol) 551.74
Formula Weight 551.74
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.