missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrrole, Thermo Scientific Chemicals

Product Code. 17211216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17211216 5 g 5g
17201216 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17211216 Supplier Thermo Scientific Acros Supplier No. 467560050
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-39-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
InChI Key YNZAFFFENDLJQG-UHFFFAOYSA-N
IUPAC Name 1H-pyrrol-1-amine
SMILES NN1C=CC=C1

Specifications

Density 1.0600 g/mL
Boiling Point 174.0°C
Quantity 5 g
Specific Gravity 1.06
Formula Weight 82.1
Chemical Name or Material 1-Aminopyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.