missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyridinium iodide, 97%

Product Code. 11491727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491727 5 g 5g
11401737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491727 Supplier Thermo Scientific Alfa Aesar Supplier No. B25703.06
  • ONLINE EXCLUSIVE PRICE
    £38.50
    £34.65 / 5g
    Save £3.85
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6295-87-0 | C5H7IN2 | 222.03 g/mol

1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

Solubility
Soluble in water (almost transparency).

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6295-87-0
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.03
MDL Number MFCD00051893
InChI Key NDRLPYIMWROJBG-UHFFFAOYSA-M
Synonym 1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay
PubChem CID 2733704
IUPAC Name pyridin-1-ium-1-amine;iodide
SMILES [I-].N[N+]1=CC=CC=C1

Specifications

Melting Point ∼160°C (decomposition)
Quantity 5 g
Beilstein 3713241
Solubility Information Soluble in water (almost transparency).
Formula Weight 222.03
Percent Purity 97%
Chemical Name or Material 1-Aminopyridinium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.