missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopiperidine, 97%

Product Code. 10037582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10037582 25 g Glass bottle 25g
10287782 10 g Glass bottle 10g
10241183 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10037582 Supplier Thermo Scientific Acros Supplier No. 104410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2213-43-6 | C5H12N2 | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2213-43-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
MDL Number MFCD00006489
InChI Key LWMPFIOTEAXAGV-UHFFFAOYSA-N
Synonym 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
PubChem CID 16658
IUPAC Name piperidin-1-amine
SMILES C1CCN(CC1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Gray or White to Yellow
Density 0.9280g/mL
Boiling Point 145.0°C to 147.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770
Quantity 25 g
Beilstein 20, 89
Specific Gravity 0.928
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with toluene and alcohol
Formula Weight 100.16
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 1-Aminopiperidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Aminopiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.