missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoindane, 99%

Product Code. 11417917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417917 1 g 1g
11427917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417917 Supplier Thermo Scientific Alfa Aesar Supplier No. A16585.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 34698-41-4 | C9H12N | 134.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34698-41-4
Molecular Formula C9H12N
Molecular Weight (g/mol) 134.20
MDL Number MFCD00003799
InChI Key XJEVHMGJSYVQBQ-VIFPVBQESA-O
Synonym 1-aminoindan, 1-aminoindane, 1-indanamine, indan-1-ylamine, indanamin, aminoindan, indanylamine, indan-1-amine, 1h-inden-1-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-1-ylamine
PubChem CID 123445
IUPAC Name 2,3-dihydro-1H-inden-1-amine
SMILES [NH3+][C@H]1CCC2=CC=CC=C12

Specifications

Melting Point 2°C
Density 1.038
Boiling Point 225°C
Flash Point 94°C (201°F)
Refractive Index 1.562
Quantity 1 g
Beilstein 3196205
Sensitivity Air Sensitive
Formula Weight 133.19
Percent Purity 99%
Chemical Name or Material (+/-)-1-Aminoindane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.