missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 10207907
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
68781-13-5
C4H7NO2·ClH
137.57
MFCD00012545
WTZRHZRMLYNBRM-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
11957445
1-aminocyclopropane-1-carboxylic acid;hydrochloride
C1CC1(C(=O)O)N.Cl
This item is not returnable. View return policy
68781-13-5
C4H7NO2·ClH
137.57
MFCD00012545
WTZRHZRMLYNBRM-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
11957445
1-aminocyclopropane-1-carboxylic acid;hydrochloride
C1CC1(C(=O)O)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 221.6°C to 225.0°C
Color White
Elemental Analysis 33.52 to 36.31% C, 5.63 to 6.09% H, 9.77 to 10.59% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 137.57
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopropanecarboxylic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Aminocyclopropanecarboxylic acid hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'