missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropanecarboxylic acid hydrochloride, 97%

Product Code. 10207767 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207767 500 mg Glass bottle 500mg
10207907 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10207767 Supplier Thermo Scientific Acros Supplier No. 429735000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68781-13-5 | C4H7NO2·ClH | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68781-13-5
Molecular Formula C4H7NO2·ClH
Molecular Weight (g/mol) 137.57
MDL Number MFCD00012545
InChI Key WTZRHZRMLYNBRM-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
PubChem CID 11957445
IUPAC Name 1-aminocyclopropane-1-carboxylic acid;hydrochloride
SMILES C1CC1(C(=O)O)N.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 221.6°C to 225.0°C
Color White
Elemental Analysis 33.52 to 36.31% C, 5.63 to 6.09% H, 9.77 to 10.59% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 137.57
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopropanecarboxylic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Aminocyclopropanecarboxylic acid hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.