missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopentanecarboxylic acid, 97+%

Product Code. 11384529
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11384529

Brand: Thermo Scientific Alfa Aesar B22179.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52-52-8 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-52-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00001381
InChI Key NILQLFBWTXNUOE-UHFFFAOYSA-N
Synonym cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
PubChem CID 2901
ChEBI CHEBI:40547
IUPAC Name 1-aminocyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)N

Specifications

Melting Point ∼320°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 636626
Merck Index 14,2734
Formula Weight 129.16
Percent Purity ≥97%
Chemical Name or Material 1-Aminocyclopentanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.