missing translation for 'onlineSavingsMsg'
Learn More

1-Aminobenzotriazole, MedChemExpress

Product Code. 16480207
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480207 50 mg 50mg
16444876 100 mg 100mg
16420976 200 mg 200mg
16411556 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480207 Supplier Medchem Express Supplier No. HY103389
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 1-Aminobenzotriazole is a nonspecific and irreversible inhibitor of cytochrome P450 (P450).

1-Aminobenzotriazole is a nonspecific and irreversible inhibitor of cytochrome P450 (P450).

Specifications

Chemical Name or Material 1-Aminobenzotriazole
Physical Form Solid
CAS 1614-12-6
Color Tan
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H6N4
For Use With (Application) Cancer-programmed cell death
Synonym ABT 3-Aminobenzotriazole
Solubility Information DMSO : 100 mg/mL (745.49 mM; Need ultrasonic) ∣H2O : 50 mg/mL (372.74 mM; Need ultrasonic)
SMILES NN1N=NC2=CC=CC=C21
Molecular Weight (g/mol) 134.14
Formula Weight 134.14
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.