missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoanthraquinone, 97%

Product Code. 11499947
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11499947

Brand: Thermo Scientific Alfa Aesar B22901.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82-45-1 | C14H9NO2 | 223.231 g/mol

In the manufacturing of dyes and pharmaceuticals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82-45-1
Melting Point 252°C to 255°C
Molecular Formula C14H9NO2
MDL Number MFCD00001213
Quantity 100 g
Beilstein 396360
Merck Index 14,417
Synonym 1-aminoanthraquinone, diazo fast red al, anthraquinone, 1-amino, 1-amino anthraquinone, 1-aminoanthrachinon, 1-amino-anthraquinone, 9,10-anthracenedione, 1-amino, alpha-aminoanthraquinone, 1-amino-9,10-anthraquinone, 1-amino-9,10-anthracenedione
Solubility Information Soluble in alcohol, benzene, chlroform, ether, glacial acetic acid, HCl
InChI Key KHUFHLFHOQVFGB-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)N
IUPAC Name 1-aminoanthracene-9,10-dione
Molecular Weight (g/mol) 223.231
PubChem CID 6710
Formula Weight 223.23
Percent Purity 97%
Chemical Name or Material 1-Aminoanthraquinone
Show More Show Less
compliance-icons
Product Identifier
  • 1-Aminoanthraquinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1-Aminoanthraquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.