missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino Hydantoin Hydrochloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1-Aminohydantoin hydrochloride |
| CAS | 2827-56-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C3 H5 N3 O2 . Cl H |
| Quantity | 1 g |
| Synonym | 2,4-Imidazolidinedione, 1-amino-, monohydrochloride (9CI), Hydantoin, 1-amino-, hydrochloride (6CI,7CI,8CI), 1-Amino-2,4-imidazolidinedione monohydrochloride, 1-Aminohydantoin hydrochloride, 1-Aminoimidazolidine-2,4-dione hydrochloride |
| InChI Formula | InChI=1S/C3H5N3O2.ClH/c4-6-1-2(7)5-3(6)8;/h1,4H2,(H,5,7,8);1H |
| SMILES | Cl.NN1CC(=O)NC1=O |
| IUPAC Name | 1-aminoimidazolidine-2,4-dione;hydrochloride |
| Molecular Weight (g/mol) | 151.55 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction