missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-9-fluorenone, 97%, Thermo Scientific™

Product Code. 11915321 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11915321 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11915321 Supplier Thermo Scientific Acros Supplier No. 103740010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6344-62-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 120.8°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C13H9NO
MDL Number MFCD00001158
Quantity 1 g
Beilstein 14,113
Synonym 1-amino-9h-fluoren-9-one, 1-amino-9-fluorenone, 9h-fluoren-9-one, 1-amino, 1-aminofluoren-9-on, acmc-1bagx, maybridge1_002094, 9h-fluoren-9-one,1-amino, 1-amino-9h-fluoren-9-one #, ksepmomkaqkosm-uhfffaoysa
InChI Key KSEPMOMKAQKOSM-UHFFFAOYSA-N
SMILES NC1=CC=CC2=C1C(=O)C1=CC=CC=C21
IUPAC Name 1-aminofluoren-9-one
Molecular Weight (g/mol) 195.22
PubChem CID 80664
Formula Weight 195.22
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-9-fluorenone
Show More Show Less
compliance-icons
Product Identifier
  • 1-Amino-9-fluorenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.