missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-hydroxyanthraquinone, 96%

Product Code. 11445827
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445827 25 g 25g
11435827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445827 Supplier Thermo Scientific Alfa Aesar Supplier No. B24388.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 116-85-8 | C14H9NO3 | 239.23 g/mol

The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

Solubility
Soluble in water, ethanol, acetone, benzene.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-85-8
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00001223
InChI Key AQXYVFBSOOBBQV-UHFFFAOYSA-N
Synonym 1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
PubChem CID 8323
IUPAC Name 1-amino-4-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N

Specifications

Melting Point 207°C to 209°C
Quantity 25 g
Solubility Information Soluble in water,ethanol,acetone,benzene.
Formula Weight 239.23
Percent Purity 96%
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.