missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-hydroxyanthraquinone 96%, Thermo Scientific™

Product Code. p-56594 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10122850

Brand: Thermo Scientific Acros 103800050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 116-85-8
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00001223
InChI Key AQXYVFBSOOBBQV-UHFFFAOYSA-N
Synonym 1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
PubChem CID 8323
IUPAC Name 1-amino-4-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 207.0°C to 209.0°C
Color Brown to Purple
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Quantity 5 g
Beilstein 14, 268
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 239.23
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Amino-4-hydroxyanthraquinone
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.