missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-cyclopentylpiperazine, 97%

Product Code. 15411008
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411008 250 mg 250mg
15490998 1 g 1g
15401008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411008 Supplier Thermo Scientific Alfa Aesar Supplier No. H54831.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61379-64-4 | C9H19N3 | 169.272 g/mol

1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61379-64-4
Molecular Formula C9H19N3
Molecular Weight (g/mol) 169.272
MDL Number MFCD00190598
InChI Key QYHRIASMJNLWHJ-UHFFFAOYSA-N
PubChem CID 1512485
IUPAC Name 4-cyclopentylpiperazin-1-amine
SMILES C1CCC(C1)N2CCN(CC2)N

Specifications

Quantity 250 mg
Synonym 1-amino-4-cyclopentylpiperazine, 1-amino-4-cyclopentyl piperazine, 4-cyclopentyl-1-piperazinamine, pubchem8581, 4-cyclopentylpiperazinylamine, 1-piperazinamine,4-cyclopentyl, 1-piperazinamine, 4-cyclopentyl, 4-cyclopentyl-piperazin-1-ylamine, 1-n-cyclopentyl piperazine-4-amine, 1-piperazinamine, 4-cyclopentyl-9ci
Solubility Information Slightly soluble in water.
Formula Weight 169.27
Percent Purity 97%
Chemical Name or Material 1-Amino-4-cyclopentylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.