missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-cyclopentylpiperazine, 97%

Product Code. 15401008
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15401008

Brand: Thermo Scientific Alfa Aesar H54831.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61379-64-4 | C9H19N3 | 169.272 g/mol

1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61379-64-4
Molecular Formula C9H19N3
Molecular Weight (g/mol) 169.272
MDL Number MFCD00190598
InChI Key QYHRIASMJNLWHJ-UHFFFAOYSA-N
PubChem CID 1512485
IUPAC Name 4-cyclopentylpiperazin-1-amine
SMILES C1CCC(C1)N2CCN(CC2)N

Specifications

Quantity 5 g
Synonym 1-amino-4-cyclopentylpiperazine, 1-amino-4-cyclopentyl piperazine, 4-cyclopentyl-1-piperazinamine, pubchem8581, 4-cyclopentylpiperazinylamine, 1-piperazinamine,4-cyclopentyl, 1-piperazinamine, 4-cyclopentyl, 4-cyclopentyl-piperazin-1-ylamine, 1-n-cyclopentyl piperazine-4-amine, 1-piperazinamine, 4-cyclopentyl-9ci
Solubility Information Slightly soluble in water.
Formula Weight 169.27
Percent Purity 97%
Chemical Name or Material 1-Amino-4-cyclopentylpiperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.