missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3-adamantanol, TRC

Product Code. 30012186
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012186 100 mg 100mg
30004159 5 g 5g
30011011 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012186 Supplier Toronto Research Chemicals Supplier No. TRCA580300100MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-82-9
Molecular Formula C10 H17 N O
Molecular Weight (g/mol) 167.25
Synonym Tricyclo[3.3.1.13,7]decan-1-ol, 3-amino-, 1-Adamantanol, 3-amino- (7CI,8CI), 3-Aminotricyclo[3.3.1.13,7]decan-1-ol, (3-Hydroxyadamantan-1-yl)amine, 1-Amino-3-adamantanol, 1-Amino-3-hydroxyadamantane, 3-Amino-1-adamantanol, 3-Amino-1-hydroxyadamantane
IUPAC Name 3-aminoadamantan-1-ol
SMILES NC12CC3CC(C1)CC(O)(C3)C2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H17NO/c11-9-2-7-1-8(3-9)5-10(12,4-7)6-9/h7-8,12H,1-6,11H2
Formula Weight 167.131
Percent Purity >95%
Chemical Name or Material 3-Aminoadamantan-1-ol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.