missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Product Code. 15476439
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476439 500 mg 500mg
15466439 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476439 Supplier Thermo Scientific Alfa Aesar Supplier No. J64527.ME
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Acetylene containing reagent with PEG spacer and carboxyl reactivity | CAS: 1013921-36-2 | C11H21NO4 | 231.292 g/mol

1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

Solubility
Soluble in DMSO, DMF, DCM, THF, or chloroform.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013921-36-2
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.292
MDL Number MFCD22380736
InChI Key QDLPAHLHHBCWOW-UHFFFAOYSA-N
Synonym 3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine
PubChem CID 86580391
IUPAC Name 2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine
SMILES C#CCOCCOCCOCCOCCN

Specifications

Color Yellow
Linear Formula NH2CH2CH2(OCH2CH2)3OCH2C≡CH
Quantity 500 mg
Solubility Information Soluble in DMSO,DMF,DCM,THF,or chloroform.
Formula Weight 231.29
Physical Form Solid
Chemical Name or Material 1-Amino-3,6,9,12-tetraoxapentadec-14-yne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.