missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,3-diethoxypropane, 97%

Product Code. 11915281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915281 100 g Glass bottle 100g
10617431 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915281 Supplier Thermo Scientific Acros Supplier No. 269851000
 Restricted Item
Estimated Shipment: 03-02-2027
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 41365-75-7 | C7H17NO2 | 147.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 41365-75-7
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
InChI Key PXXMSHBZYAOHBD-UHFFFAOYSA-N
Synonym 1-amino-3,3-diethoxypropane, 3,3-diethoxypropylamine, 3-aminopropionaldehyde diethylacetal, 1-propanamine, 3,3-diethoxy, 3-aminopropionaldehyde diethyl acetal, propionaldehyde, 3-amino-, diethyl acetal, zlchem 1268, acmc-1apws, 3,3-diethoxy-propylamine, 3,3-diethoxypropyl amine
PubChem CID 521029
IUPAC Name 3,3-diethoxypropan-1-amine
SMILES CCOC(CCN)OCC

Spécification

Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 72°C (16.0 hPa)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C2H5O)2CHCH2CH2NH2
Refractive Index 1.4233 to 1.4253
Quantity 100 g
Specific Gravity 0.91
Solubility Information Solubility in water: miscible, hydrolysis. Other solubilities: miscible with alcohol and petroleum ether
Formula Weight 147.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Amino-3, 3-diethoxypropane
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 1-Amino-3,3-diethoxypropane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Mention de danger
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.