missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylindoline, TRC

Product Code. 30340457
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340457 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340457 Supplier Toronto Research Chemicals Supplier No. TRCA634523250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-2-methylindoline
CAS 31529-46-1
Purity Grade Notes HPLC
Molecular Formula C9 H12 N2
Quantity 250 mg
Synonym 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, Indoline, 1-amino-2-methyl- (6CI,8CI), 2,3-Dihydro-2-methyl-1H-indol-1-amine, 1-Amino-2-methylindoline, N-Amino-2-methylindoline, (2RS)-2-Methyl-2,3-dihydro-1H-indol-1-amine
InChI Formula InChI=1S/C9H12N2/c1-7-6-8-4-2-3-5-9(8)11(7)10/h2-5,7H,6,10H2,1H3
SMILES CC1Cc2ccccc2N1N
IUPAC Name 2-methyl-2,3-dihydroindol-1-amine
Molecular Weight (g/mol) 148.2
Formula Weight 148.1
Percent Purity >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.