missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarboxylic acid, 99%

Product Code. 10410921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410921 250 mg Glass bottle 250mg
10617304 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10410921 Supplier Thermo Scientific Acros Supplier No. 301422500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22059-21-8 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PubChem CID 535
ChEBI CHEBI:18053
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 257°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC3H4CO2H
Quantity 250 mg
Formula Weight 101.1
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material 1-Amino-1-cyclopropanecarboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.