missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarboxylic acid, 98%

Product Code. 15416739
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416739 250 mg 250mg
15406739 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416739 Supplier Thermo Scientific Alfa Aesar Supplier No. J65118.MD
  • ONLINE EXCLUSIVE PRICE
    £56.80
    £51.65 / 250mg
    Save £5.15
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

A NMDA glutamate receptor agonist at glycine site | CAS: 22059-21-8 | C4H7NO2 | 101.11 g/mol

1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PubChem CID 535
ChEBI CHEBI:18053
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

Melting Point 229°C to 231°C
Color White
Quantity 250 mg
Beilstein 2076413
Solubility Information Soluble in water.
Formula Weight 101.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-1-cyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.