missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-1-cyclopentanecarboxylic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1-Amino-1-cyclopentanecarboxylic Acid |
| CAS | 52-52-8 |
| Molecular Formula | C6 H11 N O2 |
| Quantity | 100 mg |
| Synonym | Cyclopentanecarboxylic acid, 1-amino-, 1-Aminocyclopentanecarboxylic acid, 1-Amino-1-carboxycyclopentane, 1-Aminocyclopentane-1-carboxylic acid, 1-Azaniumylcyclopentane-1-carboxylate, ACPC, Cycloleucin, Cycloleucine, NSC 1026, NSC 112194, NSC 112195, NSC 112197, WR 14997 |
| InChI Formula | InChI=1S/C6H11NO2/c7-6(5(8)9)3-1-2-4-6/h1-4,7H2,(H,8,9) |
| SMILES | NC1(CCCC1)C(=O)O |
| IUPAC Name | 1-aminocyclopentane-1-carboxylic acid |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction