Learn More
1-Amino-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals
£69.80 - £116.00
Chemical Identifiers
CAS | 52-52-8 |
---|---|
MDL Number | MFCD00001381 |
InChI Key | NILQLFBWTXNUOE-UHFFFAOYSA-N |
Synonym | cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate |
PubChem CID | 2901 |
ChEBI | CHEBI:40547 |
IUPAC Name | 1-aminocyclopentane-1-carboxylic acid |
SMILES | C1CCC(C1)(C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10487660
|
Thermo Scientific Acros
103470050 |
5 g | Glass bottle |
£69.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10705361
|
Thermo Scientific Acros
103470100 |
10 g | Glass bottle |
£116.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52-52-8 | |
NILQLFBWTXNUOE-UHFFFAOYSA-N | |
2901 | |
1-aminocyclopentane-1-carboxylic acid |
MFCD00001381 | |
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate | |
CHEBI:40547 | |
C1CCC(C1)(C(=O)O)N |
Specifications
52-52-8 | |
100.0 | |
White to Beige | |
97% | |
H2NC5H8CO2H | |
14, I, 526 | |
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate | |
NILQLFBWTXNUOE-UHFFFAOYSA-N | |
1-aminocyclopentane-1-carboxylic acid | |
2901 | |
129.16 | |
Crystalline Flakes or Powder |
96.0 | |
320°C to 322°C | |
Authentic | |
C6H11NO2 | |
MFCD00001381 | |
15, 2727 | |
Solubility in water: ca 5g/100mL | |
C1CCC(C1)(C(=O)O)N | |
129.16 | |
CHEBI:40547 | |
97% | |
1-Amino-1-cyclopentanecarboxylic acid, 97% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-144-6
missing translation for 'rtecsNumber' : GY2625000
missing translation for 'tsca' : TSCA
- 1-Amino-1-cyclopentanecarboxylic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only