missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpiperazine, 98+%

Product Code. 11318676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318676 25 g 25g
11308676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318676 Supplier Thermo Scientific Alfa Aesar Supplier No. L19391.14
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13961-36-9 | C7H14N2 | 126.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13961-36-9
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00167970
InChI Key ZWAQJGHGPPDZSF-UHFFFAOYSA-N
Synonym 1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
PubChem CID 806422
IUPAC Name 1-prop-2-enylpiperazine
SMILES C=CCN1CCNCC1

Specifications

Density 0.902
Boiling Point 180°C to 182°C
Flash Point 51°C (123°F)
Refractive Index 1.483
Quantity 25 g
UN Number UN2920
Beilstein 507447
Sensitivity Air Sensitive
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material 1-Allylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.