missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-3-methylimidazolium bromide, 97%

Product Code. 15465858
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465858 5 g 5g
15475858 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465858 Supplier Thermo Scientific Alfa Aesar Supplier No. H59513.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31410-07-8 | C7H11BrN2 | 203.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31410-07-8
Molecular Formula C7H11BrN2
Molecular Weight (g/mol) 203.08
MDL Number MFCD08277003
InChI Key KLFDZFIZKMEUGI-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium bromide, 1-allyl-3-methyl-1h-imidazol-3-ium bromide, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium bromide, ksc221i2h, 3-methyl-1-prop-2-en-1-yl imidazol-1-ium bromide
PubChem CID 11229454
SMILES [Br-].C[N+]1=CN(CC=C)C=C1

Specifications

Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
IUPAC Name 1-methyl-3-prop-2-enylimidazol-1-ium;bromide
Formula Weight 203.08
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Allyl-3-methylimidazolium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.