missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl methyl ketone, 99%

Product Code. 10408730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10408730 25 g Glass bottle 25g
10276020 1 g Glass bottle 1g
10246070 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10408730 Supplier Thermo Scientific Acros Supplier No. 164740250
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1660-04-4 | C12H18O | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-04-4
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00074739
InChI Key DACIGVIOAFXPHW-UHFFFAOYSA-N
Synonym 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
PubChem CID 123126
IUPAC Name 1-(1-adamantyl)ethanone
SMILES CC(=O)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 55°C
Color White
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Formula Weight 178.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantyl methyl ketone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.