missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl methyl ketone, 99%

Product Code. 15442278
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442278 1 g 1g
15452278 5 g 5g
15462278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442278 Supplier Thermo Scientific Alfa Aesar Supplier No. H55889.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1660-04-4 | C12H18O | 178.28 g/mol

1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol- very faint turbidity. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-04-4
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00074739
InChI Key DACIGVIOAFXPHW-UHFFFAOYSA-N
Synonym 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
PubChem CID 123126
IUPAC Name 1-(1-adamantyl)ethanone
SMILES CC(=O)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 53°C to 55°C
Flash Point 108°C (226°F)
Quantity 1 g
Solubility Information Soluble in methanol- very faint turbidity. Insoluble in water.
Formula Weight 178.27
Percent Purity 99%
Chemical Name or Material 1-Adamantyl methyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.