missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl isocyanate, 98%

Product Code. 10531364 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10531364

Brand: Thermo Scientific Acros 314360250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4411-25-0 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4411-25-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00134405
InChI Key VBHCPGFCIQDXGZ-UHFFFAOYSA-N
Synonym 1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato
PubChem CID 2059995
IUPAC Name 1-isocyanatoadamantane
SMILES O=C=NC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 144.0°C to 146.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 177.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Adamantyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.