missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanol, 99%

Product Code. 11479532
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11479532

Brand: Thermo Scientific Alfa Aesar A10209.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-95-6 | C10H16O | 152.237 g/mol

1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-95-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00074729
InChI Key VLLNJDMHDJRNFK-UHFFFAOYSA-N
Synonym 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
PubChem CID 64152
IUPAC Name adamantan-1-ol
SMILES C1C2CC3CC1CC(C2)(C3)O

Specifications

Melting Point ∼245°C (sublimation)
Quantity 250 g
Beilstein 1903952
Solubility Information Soluble in ethanol ,methanol,and diethyl ether. Partly miscible in water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 1-Adamantanol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.