missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethylamine, 98%

Product Code. 10695612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695612 1 g Glass bottle 1g
10219163 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10695612 Supplier Thermo Scientific Acros Supplier No. 177420010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17768-41-1 | C11H20N | 166.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17768-41-1
Molecular Formula C11H20N
Molecular Weight (g/mol) 166.29
MDL Number MFCD00074750
InChI Key XSOHXMFFSKTSIT-UHFFFAOYSA-O
Synonym 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
PubChem CID 86625
IUPAC Name 1-adamantylmethanamine
SMILES [NH3+]CC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 83.0°C to 85.0°C (0.3 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5127 to 1.5147
Quantity 1 g
Specific Gravity 0.93
Formula Weight 165.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Adamantanemethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.